Skip to Content
Merck
All Photos(1)

Key Documents

P0496

Sigma-Aldrich

Anti-PTX3 antibody produced in rabbit

affinity isolated antibody, ammonium sulfate suspension

Synonym(s):

Anti-AI607804, Anti-LOC682447, Anti-PTX3, Anti-TNFAIP5, Anti-TSG-14, Anti-pentraxin 3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
WB
clone:
polyclonal
species reactivity:
rodent, human
citations:
6
technique(s):
immunohistochemistry: suitable
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

ammonium sulfate suspension

mol wt

antigen 31.8 kDa

species reactivity

rodent, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

storage temp.

−20°C

Gene Information

General description

The PTX3 (pentraxin 3) gene is mapped to human chromosome 3q25.32. The protein is localized in mesenchymal cells, endothelial cells of vascular tissue, fibroblasts, monocytes and granulocytes.

Immunogen

a synthetic peptide corresponding to human PTX3 (amino acids 110-124) conjugated to BSA.

Application

Anti-PTX3 antibody produced in rabbit has been used in immunofluorescence and immunohistochemistry.

Biochem/physiol Actions

PTX3 (pentraxin 3) is associated with humoral immune response and inflammatory processes, and therefore offers protection against pathogenic antigens. Proinflammatory cytokines, microbes and signals derived from damaged tissue triggers the expression of the gene upon stimulation. It serves as a biomarker for a number of inflammatory vascular and autoimmune disorders, including vasculitis, rheumatoid arthritis, psoriasis, and cardiovascular diseases. PTX3 is involved in tumor progression by participating in proliferation and invasion steps, which were observed in glioma.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of the association of plasma pentraxin 3 levels with type 2 diabetes and diabetic nephropathy in a Malay population.
Abu Seman N, et al.
Journal of Diabetes Research, 2013 (2013)
Long pentraxin 3: a marker of inflammation in untreated psoriatic patients.
Bevelacqua V, et al.
International Journal of Molecular Medicine, 18(3), 415-423 (2006)
Growth arrest specific-1 (GAS1) is a C/EBP target gene that functions in ovulation and corpus luteum formation in mice.
Ren Y A, et al.
Biology of Reproduction, 94(2), 44-41 (2016)
Inhibition of pentraxin 3 in glioma cells impairs proliferation and invasion in vitro and in vivo.
Tung J N, et al.
Journal of Neuro-Oncology, 129(2), 201-209 (2016)
Pentraxin 3 plasma levels at graft-versus-host disease onset predict disease severity and response to therapy in children given haematopoietic stem cell transplantation.
Dander E, et al.
Oncotarget, 7(50), 82123-82123 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service