Skip to Content
Merck
All Photos(2)

Key Documents

MAK210

Sigma-Aldrich

Human Calpain 1 Inhibitor Screening Kit

Sufficient for 100 Fluorometric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.25

usage

sufficient for 100 fluorometric tests

detection method

fluorometric

relevant disease(s)

cardiovascular diseases; cancer

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CAPN1(823)

General description

Mammalian calpains are calcium-activated, neutral, non-lysosomal, intracellular proteases that may be ubiquitous, stomach-specific, or muscle-specific. Calpain 1 is a ubiquitous protease that is involved in calcium homeostasis and the regulation of apoptosis. In cardiomyocytes, activated calpain 1 induces apoptosis by BID cleavage and release of cytochrome c from the mitochondria.

The Human Calpain 1 Inhibitor Screening Kit is a rapid, simple and sensitive assay that is suitable for high-throughput screening of Calpain 1 inhibitors. Calpain 1 activity is measured by cleaving a synthetic 7-amino-4-trifluoromethylcoumarin (AFC)-based substrate (Ac-LLY-AFC) to yield AFC, a fluorescent product (λex = 400/λem = 505 nm), proportional to the enzymatic activity present.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

Suitable for the screening of inhibitors of human Calpain 1.

Principle

The Human Calpain 1 Inhibitor Screening Kit is a rapid, simple and sensitive assay that is suitable for high-throughput screening of Calpain 1 inhibitors. Calpain 1 activity is measured by cleaving a synthetic 7-amino-4-trifluoromethylcoumarin (AFC)-based substrate (Ac-LLY-AFC) to yield AFC, a fluorescent product (λex = 400/λem = 505 nm), proportional to the enzymatic activity present.

Kit Components Only

Product No.
Description

  • Assay Buffer

  • Substrate

  • Calpain 1 Enzyme

  • Inhibitor Control

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service