L7655
Latex beads, carboxylate-modified, biotin-labeled, blue
buffered aqueous suspension, 0.25 μm mean particle size
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
buffered aqueous suspension
composition
Solids, 1.0%
mean particle size
0.25 μm
Looking for similar products? Visit Product Comparison Guide
Application
Carboxylate-modified polystyrene latex beads have been used to collect morphofunctional data about the immune systems of Carabus lefebvrei, a helicophagous Italian endemic ground beetle often used as an indicator of the habitat quality of the Apennines mountain forests. Carboxylate-modified polystyrene latex beads have also been used to inform comparative studies on immunity defense mechanisms of adults and larvae of the coleopteran Cetonischema aeruginosa and to investigate the phagocytic activity of cultured rat thyroid cells.
Physical form
Suspension in 10 mM PBS, pH 7.1, 2 mM NaN3
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Ronen Adato et al.
Nature communications, 4, 2154-2154 (2013-07-24)
Infrared absorption spectroscopy is a powerful biochemical analysis tool as it extracts detailed molecular structural information in a label-free fashion. Its molecular specificity renders the technique sensitive to the subtle conformational changes exhibited by proteins in response to a variety
K Takazawa et al.
Acta endocrinologica, 117(2), 198-204 (1988-02-01)
We investigated the phagocytic activity of cultured rat thyroid cells (FRTL) by using latex beads. FRTL cells ingested many carboxylate-modified polystyrene latex beads (c-latex beads), which had carboxyl residues on the surface, whereas they ingested few polystyrene latex beads (p-latex
P G Giulianini et al.
Tissue & cell, 35(4), 243-251 (2003-08-19)
In the context of comparative studies on immunity defence mechanisms of adults and larvae of the coleopteran Cetonischema aeruginosa (Drury, 1770) the ultrastructure of the circulating hemocytes of the third instar larval stage has been investigated by means of light
Circulating hemocytes from larvae and adults of Carabus (Chaetocarabus) lefebvrei Dejean 1826 (Coleoptera, Carabidae): cell types and their role in phagocytosis after in vivo artificial non-self-challenge
Giglio, A., et al.
Micron (Oxford, England : 1993), 39, 52-58 (2008)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service