J3774
Monoclonal Anti-JAK1 antibody produced in mouse
2 mg/mL, clone JAK1-193, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-Janus Kinase 1
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
JAK1-193, monoclonal
form
buffered aqueous solution
mol wt
antigen 130 kDa
species reactivity
rat, human, mouse
concentration
2 mg/mL
technique(s)
indirect ELISA: suitable
microarray: suitable
western blot: 2-4 μg/mL using extract from Raji cells
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... JAK1(3716)
mouse ... Jak1(16451)
rat ... Jak1(84598)
General description
Immunogen
Application
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service