G112050
DMT-dG(tac) Phosphoramidite
configured for ABI, configured for (ÄKTA® and OligoPilot®)
Synonym(s):
DMT-dG(tac) amidite
About This Item
type
for DNA synthesis
product line
Proligo Reagents
Assay
≥98% (31P-NMR)
≥98.0% (reversed phase HPLC)
technique(s)
oligo synthesis: suitable
λ
conforms (UV/VIS Identity)
compatibility
configured for (ÄKTA® and OligoPilot®)
configured for ABI
nucleoside profile
base: deoxyguanosine
base protecting group: TAC
2' protecting group: none
5' protecting group: DMT
deprotection: fast
storage temp.
-10 to -25°C
SMILES string
COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n3cnc4C(=O)NC(NC(=O)COc5ccc(cc5)C(C)(C)C)=Nc34)(c6ccccc6)c7ccc(OC)cc7
InChI
1S/C52H62N7O9P/c1-34(2)59(35(3)4)69(66-29-13-28-53)68-43-30-46(58-33-54-47-48(58)56-50(57-49(47)61)55-45(60)32-64-42-26-16-36(17-27-42)51(5,6)7)67-44(43)31-65-52(37-14-11-10-12-15-37,38-18-22-40(62-8)23-19-38)39-20-24-41(63-9)25-21-39/h10-12,14-27,33-35,43-44,46H,13,29-32H2,1-9H3,(H2,55,56,57,60,61)/t43-,44+,46+,69?/m0/s1
InChI key
MCBSUBQIVLOCIW-YEZUPXOUSA-N
Looking for similar products? Visit Product Comparison Guide
General description
- Deprotection of the TAC group is ultra-fast: complete deprotection inconcentrated ammonia occurs within 15 minutes at 55 °C or two hours atroom temperature
- Compatible with the AMA deprotection reagent (a mixture of ≥25%ammonia in water with 40% aqueous methylamine I/I, v/v)
- Highly soluble in acetonitrile. No need to add co-solvents such asdimethylformamide or methylene chloride
- Suitable for the synthesis of oligomers with base-labile units e.g., dyes andmodifiers, because of less exposure to ammonia and the possibility ofroom temperature deprotection
- No change is required in the reagents commonly used for DNA synthesis,except that Proligo′s Fast Deprotection Cap A solution is used instead ofCap A solution
- The application of dA(tac) minimizes depurination and improves thequality of oligonucleotidesChanging the dC protecting group to the TAC protecting group leads torapid synthesis of high-purity and high-yield oligonucleotides. Substitutingthe commonly employed dC(bz) monomer by the dC(tac) monomerenables the application of ultrafast deprotection with the AMA reagent andprovides a high-throughput method of oligonucleotide synthesis.DMT-dC(tac) Phosphoramidite configured ABI Synthesizers.
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service