Skip to Content
Merck
All Photos(1)

Key Documents

E0281

Sigma-Aldrich

Anti-Estrogen-Related Receptor γ antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ERRγ, Anti-NR3B3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC (p)
clone:
polyclonal
species reactivity:
human
citations:
2
technique(s):
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 17 μg/mL using human breast tissue

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

availability

not available in Japan

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 17 μg/mL using human breast tissue

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ESRRG(2104)
mouse ... Esrrg(26381)
rat ... Esrrg(360896)

Immunogen

synthetic peptide corresponding to the internal domain of human estrogen-related receptor γ conjugated to KLH. The immunizing peptide has 100% homology with the rat and mouse gene.

Biochem/physiol Actions

Estrogen-Related Receptors (ERR) are hormone receptors that bind estrogen response elements that in turn bind specific DNA sequences and regulate transcription. Three ERRs are known, alpha, beta and gamma, with highly conserved ligand and DNA binding sequences. ERRγ is selectively expressed in brain and is an important modulator of energy balance, food intake, neuropeptide levels and response to stress.

Physical form

Solution in phosphate buffered saline, pH 7.7, containing 0.01% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William A Alaynick
Mitochondrion, 8(4), 329-337 (2008-04-01)
Lipid metabolism is a continuum from emulsification and uptake of lipids in the intestine to cellular uptake and transport to compartments such as mitochondria. Whether fats are shuttled into lipid droplets in adipose tissue or oxidized in mitochondria and peroxisomes
Mardi S Byerly et al.
BMC physiology, 13, 10-10 (2013-09-24)
Estrogen-related receptors (ERRs) are orphan nuclear hormone receptors expressed in metabolically active tissues and modulate numerous homeostatic processes. ERRs do not bind the ligand estrogen, but they are able to bind the estrogen response element (ERE) embedded within the ERR
Damian Ryszawy et al.
Toxicology in vitro : an international journal published in association with BIBRA, 62, 104676-104676 (2019-10-21)
Bisphenol A (BPA) is commonly present in plastics used for food storage and preservation. The release of BPA from these products results in a permanent human exposition to BPA; however, the quality and quantity of BPA adverse effects remain a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service