Skip to Content
Merck
All Photos(1)

Key Documents

94967

Sigma-Aldrich

2-cis,6-trans-Farnesol

≥95.0% (GC)

Synonym(s):

(2Z,6E)-3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol, cis,trans-Farnesol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26O
CAS Number:
Molecular Weight:
222.37
Beilstein:
1723038
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95.0% (GC)

form

liquid

storage temp.

−20°C

SMILES string

C\C(C)=C\CC\C(C)=C\CC\C(C)=C/CO

InChI

1S/C15H26O/c1-13(2)7-5-8-14(3)9-6-10-15(4)11-12-16/h7,9,11,16H,5-6,8,10,12H2,1-4H3/b14-9+,15-11-

InChI key

CRDAMVZIKSXKFV-PVMFERMNSA-N

Biochem/physiol Actions

Farnesol is an acyclic sesquiterpene alcohol, with four possible isomers, the E/E-isomer being the most common in nature. It was found to have different biological functions, for example to act as a quorum-sensing molecule to suppress filamentation in the fungus Candida albicans; and to have antibacterial effects against Staphylococcus aureus, including promotion of potassium ion leakage. Its derivatives are involved in the a-factor mating peptide of the dodecapeptide pheromone found in Saccharomyces cerevisiae. The biological activity of farnesol and FPP analogues are investigated in cancer research. 2-cis,6-trans-Farnesol is a metabolite in the sesquiterpenoid and triterpenoid biosynthesis, a reactant of farnesol 2-isomerase (EC: 5.2.1.9)

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Xie et al.
The Journal of organic chemistry, 65(25), 8552-8563 (2000-12-12)
The a-factor of Saccharomyces cerevisiae is a dodecapeptide pheromone (YIIKGVFWDPAC(Farnesyl)-OCH(3), 1), in which post-translational modification with a farnesyl isoprenoid and carboxymethyl group is required for full biological activity. This peptide has been used as a model system to explore the
Yoshihiro Inoue et al.
FEMS microbiology letters, 237(2), 325-331 (2004-08-24)
The study was made of the antibacterial effects of three terpene alcohols on Staphylococcus aureus, focusing on the leakage of K+ ions and toxicity over time. The leakage of K+ ions was monitored continuously with a K+-electrode. Our results suggested
R J Hohl et al.
Lipids, 33(1), 39-46 (1998-02-21)
This investigation compares the effects of three farnesyl pyrophosphate analogs on selected aspects of isoprenoid metabolism. E,E-alpha-Hydroxyfarnesylphosphonate was prepared by an improved variation on a literature synthesis, which also gave access to the new Z,E-alpha-hydroxyfarnesyl- and alpha-hydroxygeranylphosphonates. A striking find
Roman Shchepin et al.
Chemistry & biology, 10(8), 743-750 (2003-09-05)
The dimorphic fungus Candida albicans produces extracellular farnesol (3,7,11-trimethyl-2,6,10-dodecatriene-1-ol) which acts as a quorum-sensing molecule (QSM) to suppress filamentation. Of four possible geometric isomers of farnesol, only the E,E isomer possesses QSM activity. We tested 40 natural and synthetic analogs
J Downward
Current opinion in genetics & development, 8(1), 49-54 (1998-04-08)
Activated Ras proteins have either positive or negative effects on the regulation of apoptosis depending on cell type and other factors. In part, this is due to the ability of Ras to control directly multiple effector pathways, including PI3-kinase, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service