Skip to Content
Merck
All Photos(1)

Key Documents

82348

Sigma-Aldrich

Lithium sulfate solution

BioUltra, 2 M in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Li2O4S
CAS Number:
Molecular Weight:
109.94
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25
form:
liquid

product line

BioUltra

form

liquid

concentration

2 M in H2O

impurities

insoluble matter, passes filter test

pH

6.0-9.0 (25 °C, 2 M in H2O)

density

1.168 g/mL at 20 °C

anion traces

chloride (Cl-): ≤50 mg/kg
nitrate (NO3-): ≤10 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤10 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

2 M in H2O

UV absorption

λ: 260 nm Amax: 0.08
λ: 280 nm Amax: 0.04

SMILES string

[Li+].[Li+].[O-]S([O-])(=O)=O

InChI

1S/2Li.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

INHCSSUBVCNVSK-UHFFFAOYSA-L

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuya Yamada et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 62(Pt 10), 1016-1017 (2006-10-03)
Atg3 is an E2-like enzyme that catalyzes the conjugation reaction between Atg8 and phosphatidylethanolamine (PE). The Atg8-PE conjugate is essential for autophagy, the bulk degradation process of cytoplasmic components by the vacuolar/lysosomal system. Crystals of Saccharomyces cerevisiae Atg3 have been
Sanjay Kasture et al.
Natural product research, 25(20), 1950-1954 (2011-06-29)
In the traditional system of medicine in Sardinia, Rubia peregrina (RP) is reported as an aphrodisiac herb. Since aphrodisiacs may also have dopaminergic activity, and there can be a reciprocal relationship between dopaminergic and serotonergic functions in the central nervous
O I Volkov
Zhurnal obshchei biologii, 71(4), 359-368 (2010-09-28)
The data from scientific publications on excretory activity of herbs root endings were analyzed, along with the data on the role of polyvalent metals cations in stabilization of humus substances (HS) of soil organic mineral complex. On the base of
Benedikt Habermeyer et al.
Psychiatrische Praxis, 35(4), 198-200 (2007-11-08)
Lithium is known to be neurotoxic at higher serum levels. In rare cases patients develop symptoms of intoxication or full blown intoxications even when lithium levels are normal. Our aim was to detect predicting factors of lithium neurotoxicity at normal
Janaína Teles de Faria et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 877(27), 3031-3037 (2009-09-01)
Xylose reductase (XR) from Debaryomyces hansenii was extracted by partitioning in aqueous two-phase systems (ATPS) composed of polyethylene glycol (PEG) 4000 in the presence of different salts, specifically sodium sulfate, lithium sulfate and potassium phosphate. Batch extractions were carried out

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service