Skip to Content
Merck
All Photos(1)

Key Documents

53609

Sigma-Aldrich

Robinin

≥90.0% (HPLC)

Synonym(s):

Kaempferol 3-O-robinoside-7-O-rhamnoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H40O19
CAS Number:
Molecular Weight:
740.66
EC Number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Assay

≥90.0% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(Oc4cc(O[C@@H]5O[C@@H](C)[C@H](O)[C@@H](O)[C@H]5O)cc(O)c4C3=O)c6ccc(O)cc6)[C@H](O)[C@@H](O)[C@H]2O)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C33H40O19/c1-10-19(36)23(40)26(43)31(47-10)46-9-17-21(38)25(42)28(45)33(51-17)52-30-22(39)18-15(35)7-14(49-32-27(44)24(41)20(37)11(2)48-32)8-16(18)50-29(30)12-3-5-13(34)6-4-12/h3-8,10-11,17,19-21,23-28,31-38,40-45H,9H2,1-2H3/t10-,11-,17+,19-,20-,21-,23+,24+,25-,26+,27+,28+,31+,32-,33-/m0/s1

InChI key

PEFASEPMJYRQBW-HKWQTAEVSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Flavone glycoside from Robinia with antibacterial and diuretic properties; derived from kaempferol.
The ability of robinin to reverse the multidrug resistance of the human colon cancer cell line Colo 320 expreβing MDR1/LRP was explored by monitoring their uptake of Rhodamine 123, but was shown to have minimal effect. Robinin was detected in lysates of the human breast cancer cell line MCF-7 using liquid chromatography-tandem maβ spectrometry.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Robinin intoxication].
B Lagarón Cabello et al.
Atencion primaria, 30(4), 258-259 (2002-09-19)
Raymond E March et al.
Rapid communications in mass spectrometry : RCM, 18(9), 931-934 (2004-04-30)
A mass spectrometric method based on the combined use of electrospray ionization, collision-induced dissociation and tandem mass spectrometry has been applied to the structural characterization of the flavone triglycoside, robinin (3,5,7,4'-tetrahydroxyflavone-3-O-robinoside-7-O-rhamnoside). The deprotonated molecule fragments by loss of the rhamnose
Lia Tsiklauri et al.
Journal of pharmaceutical and biomedical analysis, 55(1), 109-113 (2011-01-15)
An accurate, precise and sensitive method was developed and validated for the simultaneous quantification of the flavonoid glycoside robinin, and its algycone kaempferol in human breast cancer MCF-7 cells. The application of liquid chromatography-tandem mass spectrometry (LC/MS/MS) with a TurboIonspray
V D Bokkenheuser et al.
The Biochemical journal, 248(3), 953-956 (1987-12-15)
Rutin and quercitrin are hydrolysed to quercetin, and robinin is hydrolysed to kaempferol, by faecal flora from healthy subjects. The enzymes required for these hydrolyses, namely alpha-rhamnosidase and beta-galactosidase, were produced by some strains of Bacteroides distasonis; other strains, however
Roles of flavonoids in plant resistance to insects.
P A Hedin et al.
Progress in clinical and biological research, 213, 87-100 (1986-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service