Skip to Content
Merck
All Photos(1)

Key Documents

50984

Sigma-Aldrich

1,2-Dipalmitoyl-sn-glycero-3-phospho-rac-(1-glycerol) sodium salt

≥96.0% (TLC)

Synonym(s):

1,2-Dihexadecanoyl-sn-glycero-3-phospho-(1′-rac-glycerol) sodium salt, L-α-Phosphatidyl-DL-glycerol, dipalmitoyl, DPPG-Na, Hexadecanoic acid 1-[[[(2,3-dihydroxypropoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl ester, PG(16:0/16:0)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C38H74O10PNa
CAS Number:
Molecular Weight:
744.95
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

biological source

synthetic

Assay

≥96.0% (TLC)

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCC(O)CO)=O)(OC(CCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCC)=O.[Na+]

InChI

1S/C38H75O10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-37(41)45-33-36(34-47-49(43,44)46-32-35(40)31-39)48-38(42)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h35-36,39-40H,3-34H2,1-2H3,(H,43,44);/q;+1/p-1/t35?,36-;/m1./s1

InChI key

LDWIWSHBGAIIMV-ODZMYOIVSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Forms classical lipid bilayers. At low temperature, however, an unusual and much thinner bilayer forms where the hydrocarbon chains are interdigitated.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

PG(16:0/16:0)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service