33407
2-Butanone
puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Synonym(s):
Ethyl methyl ketone, MEK, Methyl ethyl ketone
About This Item
Recommended Products
grade
ACS reagent
puriss. p.a.
Quality Level
Agency
reag. Ph. Eur.
vapor density
2.49 (vs air)
vapor pressure
71 mmHg ( 20 °C)
Assay
≥99.5% (GC)
form
liquid
autoignition temp.
960 °F
expl. lim.
10.1 %
impurities
≤0.0003% KMnO4 red. matter (as O)
≤0.002% non-volatile matter
≤0.003% free acid (as CH3COOH)
≤0.05% 2-butanol
≤0.05% water (Karl Fischer)
color
APHA: <15
refractive index
n20/D 1.3770-1.3810
n20/D 1.379 (lit.)
bp
80 °C (lit.)
mp
−87 °C (lit.)
density
0.805 g/mL at 25 °C (lit.)
cation traces
Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sn: ≤0.1 mg/kg
Zn: ≤0.2 mg/kg
SMILES string
CC(CC)=O
InChI
1S/C4H8O/c1-3-4(2)5/h3H2,1-2H3
InChI key
ZWEHNKRNPOVVGH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
26.6 °F - closed cup
Flash Point(C)
-3 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service