Skip to Content
Merck
All Photos(1)

Key Documents

B0100000

Bacitracin zinc

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Bacitracin zinc salt, Zinc bacitracin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C66H101N17O16SZn
CAS Number:
Molecular Weight:
1486.07
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

API family

bacitracin

mp

250 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Zn++].CCC(C)C(N)C1=NCC(S1)C(=O)N[C@@H](CC(C)C)C(=O)N[C@H](CCC([O-])=O)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@H]2CCCCNC(=O)[C@H](CC(N)=O)NC(=O)[C@@H](CC([O-])=O)NC(=O)[C@H](Cc3cnc[nH]3)NC(=O)[C@@H](Cc4ccccc4)NC(=O)[C@@H](NC(=O)[C@@H](CCCN)NC2=O)[C@@H](C)CC

InChI

1S/C66H103N17O16S.Zn/c1-9-35(6)52(69)66-72-32-48(100-66)63(97)80-43(26-34(4)5)59(93)75-42(22-23-50(85)86)58(92)83-53(36(7)10-2)64(98)76-40-20-15-16-25-71-55(89)46(29-49(68)84)78-62(96)47(30-51(87)88)79-61(95)45(28-39-31-70-33-73-39)77-60(94)44(27-38-18-13-12-14-19-38)81-65(99)54(37(8)11-3)82-57(91)41(21-17-24-67)74-56(40)90;/h12-14,18-19,31,33-37,40-48,52-54H,9-11,15-17,20-30,32,67,69H2,1-8H3,(H2,68,84)(H,70,73)(H,71,89)(H,74,90)(H,75,93)(H,76,98)(H,77,94)(H,78,96)(H,79,95)(H,80,97)(H,81,99)(H,82,91)(H,83,92)(H,85,86)(H,87,88);/q;+2/p-2/t35?,36-,37-,40-,41+,42+,43-,44+,45-,46-,47+,48?,52?,53-,54-;/m0./s1

InChI key

NKULICGUDKGGRL-FCHFGNCGSA-L

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Bacitracin zinc EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soo Hyun Lee et al.
Biopharmaceutics & drug disposition, 33(6), 292-303 (2012-07-19)
To evaluate the metabolic interaction between host and gut microflora on drug metabolism, pseudo germ-free rats were prepared with an antibiotics cocktail to change their gut conditions. The usefulness of the pseudo germ-free model was evaluated for observing the DMPK
Shanna Spring et al.
Dermatitis : contact, atopic, occupational, drug, 23(5), 210-213 (2012-09-27)
Topical medicaments are a common cause of allergic contact dermatitis. This study will evaluate the prevalence of contact allergy to a wide array of topical medicaments at the Ottawa Patch Test Clinic. The objectives of this study are to report
Audrey Charlebois et al.
PloS one, 7(9), e44449-e44449 (2012-09-13)
Phenotypic bacitracin resistance has been reported in Clostridium perfringens. However, the genes responsible for the resistance have not yet been characterized. Ninety-nine C. perfringens isolates recovered from broilers and turkeys were tested for phenotypic bacitracin resistance. Bacitracin MIC(90) (>256 µg/ml)
David B Adimpong et al.
Applied and environmental microbiology, 78(22), 7903-7914 (2012-09-04)
Bacillus spp. are widely used as feed additives and probiotics. However, there is limited information on their resistance to various antibiotics, and there is a growing concern over the transfer of antibiotic resistance genes. The MIC for 8 antibiotics was
Marta Estela Saravia et al.
Archives of oral biology, 58(3), 311-316 (2012-11-15)
The recovery of mutans streptococci in saliva and dental biofilm samples depends, in part, on the culture medium used. In this study, we compared (i) the culture media Sucrose-Bacitracin agar (SB-20), Modified SB-20 (SB-20M) and Mitis Salivarius Bacitracin agar (MSB)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service