Skip to Content
Merck
All Photos(1)

Key Documents

80479

Supelco

Bergenin

analytical standard

Synonym(s):

Ardisic acid B, Cuscutin, Vakerin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16O9
CAS Number:
Molecular Weight:
328.27
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

InChI

1S/C14H16O9/c1-21-11-5(16)2-4-7(9(11)18)12-13(23-14(4)20)10(19)8(17)6(3-15)22-12/h2,6,8,10,12-13,15-19H,3H2,1H3/t6-,8-,10+,12+,13-/m1/s1

InChI key

YWJXCIXBAKGUKZ-HJJNZUOJSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Birger Dittrich et al.
Acta crystallographica. Section B, Structural science, 65(Pt 6), 749-756 (2009-11-20)
Bergenin, which has been isolated from a variety of tropical plants, has several pharmacological applications in traditional Asian medicine. Its electron-density distribution was obtained from a room-temperature low-resolution X-ray data set measured with point detection making use of multipole populations
Weisong Yu et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 877(1-2), 33-36 (2008-11-28)
This paper reports the development and validation of an assay for quantitation of bergenin in human plasma using liquid chromatography/tandem mass spectrometry (LC-MS/MS). Bergenin and the internal standard (I.S.), 5-bromo-2,4(1H,3H)-pyrimidinedione (5-BrU), were separated by reversed phase HPLC and quantitated by
Nighat Nazir et al.
European journal of medicinal chemistry, 46(6), 2415-2420 (2011-04-09)
Bergenin pentacetate (2), a peracetate derivative of biologically active lead compound Bergenin (1) isolated from Bergenia stracheyi was subjected to lipase catalyzed regioselective alcoholysis to obtain 3,4,10,11-tetracetate of Bergenin (3). The free hydroxyl group of 3 was derivatised using higher
Haojing Song et al.
Biomedical chromatography : BMC, 27(11), 1398-1405 (2013-05-23)
Bergenin is the major component of Ardisia creanta sims and Rodgersia sambucifolia hemsl with many biological activities. Although bergenin has been used to treat human diseases in China for man years, there is no report regarding its metabolism. This is
Muhammad Raza Shah et al.
Bioorganic & medicinal chemistry letters, 22(8), 2744-2747 (2012-03-23)
Bergenin is an isocoumarin natural product which aides in fat loss, healthy weight maintenance, enhancing the lipolytic effects of norepinephrine, inhibiting the formation of interleukin 1α and cyclooxygenases-2. Here we describe the anti-inflammatory activity of new bergenin derivatives 1-15 in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service