Skip to Content
Merck
All Photos(1)

Key Documents

76466

Supelco

(−)-Sparteine

≥98.0% (GC)

Synonym(s):

(−)-Lupinidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26N2
CAS Number:
Molecular Weight:
234.38
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (GC)

form

liquid

optical activity

[α]/D -17.5±1.5°, c = 1 in ethanol

refractive index

n20/D 1.526-1.530
n20/D 1.528 (lit.)

bp

137-138 °C/1 mmHg (lit.)

density

1.02 g/mL at 25 °C (lit.)

SMILES string

C1CCN2C[C@@H]3C[C@@H](CN4CCCC[C@H]34)[C@@H]2C1

InChI

1S/C15H26N2/c1-3-7-16-11-13-9-12(14(16)5-1)10-17-8-4-2-6-15(13)17/h12-15H,1-11H2/t12-,13-,14-,15+/m0/s1

InChI key

SLRCCWJSBJZJBV-ZQDZILKHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sparteine belongs to lupin alkaloid class of compounds. It is a well-known neurological toxin.

Application

(−)-Sparteine may be used as a chiral diamino ligand in the asymmetric catalysis reactions involving alkyllithium bases. It may be used as a reference standard for the determination of (−)-sparteine in animal-derived food products by modified quick, easy, cheap, effective, rugged, and safe (CEN QuEChERS) extraction method followed by analysis using liquid chromatography-electrospray ionization tandem mass spectrometry (LC-ESI/MS-MS).

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New chiral diamino ligands as sparteine analogues. Application to the palladium-catalyzed kinetic oxidative resolution of 1-phenyl ethanol.
Lesma G, et al.
Tetrahedron Asymmetry, 19(11), 1363-1366 (2008)
A modified QuEChERS method coupled with liquid chromatography-tandem mass spectrometry for the simultaneous detection and quantification of scopolamine, L-hyoscyamine, and sparteine residues in animal-derived food products.
Zheng W, et al.
Journal of Advanced Research (2018)
Microwave-assisted extraction of tanshinones from Salvia miltiorrhiza bunge with analysis by high-performance liquid chromatography.
Pan X, et al.
Journal of Chromatography A, 922(1-2), 371-375 (2001)
Sparteine as a chiral ligand for asymmetric catalysis
Chiral Diazaligands for Asymmetric Synthesis, 59-92 (2005)
Diurnal cycle of sparteine production in Lupinus arcticus.
Sharam GJ and Turkington R
Canadian Journal of Botany. Journal Canadien De Botanique, 83(10), 1345-1348 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service