Skip to Content
Merck
All Photos(1)

Key Documents

68291

Supelco

Ethyl salicylate

analytical standard

Synonym(s):

Ethyl 2-hydroxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-(HO)C6H4CO2C2H5
CAS Number:
Molecular Weight:
166.17
Beilstein:
907659
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

0.05 mmHg ( 25 °C)

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.522 (lit.)
n20/D 1.522-1.524

bp

234 °C (lit.)

mp

1 °C (lit.)

density

1.131 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CCOC(=O)c1ccccc1O

InChI

1S/C9H10O3/c1-2-12-9(11)7-5-3-4-6-8(7)10/h3-6,10H,2H2,1H3

InChI key

GYCKQBWUSACYIF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl salicylate belongs to the class of terpenoids. It is one of the flavor additives typically used in tobacco products. Ethyl salicylate also finds applications as a fragrance ingredient in perfumes, decorative cosmetics, fine fragrances, shampoos, and detergents.

Application

Ethyl salicylate may be used as an analytical standard for the determination of the analyte in biological fluids and topical formulation by chromatography-based techniques.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fragrance material review on ethyl salicylate
Lapczynski A, et al.
Food And Chemical Toxicology, 45(1), S397-S401 (2007)
Wound healing activity of aqueous extracts of leaves and roots of Coleus aromaticus in rats
Jain AK<, et al.
Acta Poloniae Pharmaceutica, 69, 1119-1123 (2012)
T Kakkar et al.
Journal of chromatography. B, Biomedical sciences and applications, 718(1), 69-75 (1998-12-01)
A gas chromatographic-mass spectrometric (GC-MS) assay was developed for the quantitative analysis of methyl salicylate (MeS), ethyl salicylate (ES) and salicylic acid (SA) from biological fluids. The method was validated from 100-microl rat liver homogenate preparations (5 mg/ml protein) in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service