Skip to Content
Merck
All Photos(1)

Key Documents

63178

Sigma-Aldrich

(4-Maleimidobutyramidomethyl)polystyrene

extent of labeling: ~0.4 mmol/g "maleimide" loading

Synonym(s):

‘Maleimide’ polymer-bound, ‘Polymer-supported maleimide’

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

extent of labeling

~0.4 mmol/g "maleimide" loading

matrix

crosslinked with 1% DVB

particle size

200-400 mesh

storage temp.

2-30°C

Application

Polymer(polystyrene)-supported maleimide ((4-Maleimidobutyramidomethyl)polystyrene)) may be use for solid phase trapping of thiols from solutions and for the immobilization of sulhydryl-containing substances such as proteins and peptides.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M F Lazo-Javalera et al.
SpringerPlus, 5, 453-453 (2016-04-28)
Establishment of an efficient explants surface disinfection protocol is essential for in vitro cell and tissue culture as well as germplasm conservation, such as the case of Grapevine (Vitis spp.) culture. In this research, different procedures for disinfection and regeneration
Ksenia A Sedova et al.
International journal of molecular sciences, 20(24) (2019-12-19)
Melatonin is assumed to confer cardioprotective action via antioxidative properties. We evaluated the association between ventricular tachycardia and/or ventricular fibrillation (VT/VF) incidence, oxidative stress, and myocardial electrophysiological parameters in experimental ischemia/reperfusion under melatonin treatment. Melatonin was given to 28 rats
Engin Yıldırım et al.
Anadolu kardiyoloji dergisi : AKD = the Anatolian journal of cardiology, 11(4), 343-350 (2011-05-13)
Although estrogens have been shown to be vasoactive hormones, the vascular effects of testosterone are not well defined. Little is known about the in vitro effects of testosterone on isolated arteries. The purpose of this experimental prospective study was to
Min Zhong et al.
Plant science : an international journal of experimental plant biology, 296, 110492-110492 (2020-06-17)
Transglutaminases (TGases), mediators of the transamidation of specific proteins by polyamines (PA), play critical roles in PA metabolism in animals, but their functions and regulatory mechanisms are largely unknown in plants. In this study, we demonstrated that TGase from cucumber
Bernard J Moncla et al.
PloS one, 11(7), e0158687-e0158687 (2016-07-22)
In this study, we characterized the glycome of cervical-vaginal fluid, collected with a Catamenial cup. We quantified: glycosidase levels; sialic acid and high mannose specific lectin binding; mucins, MUC1, MUC4, MUC5AC, MUC7; and albumin in the samples collected. These data

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service