611913
Potassium fluoride solution
NMR reference standard, 0.2 M in water, D2O 10 %, NMR tube size 10 mm × 8 in.
About This Item
Recommended Products
grade
NMR reference standard
analytical standard
Quality Level
composition
D2O, 10%
concentration
0.2 M in water
technique(s)
NMR: suitable
NMR tube size
10 mm × 8 in.
suitability
suitable for NMR (reference standard)
format
single component solution
SMILES string
[F-].[K+]
InChI
1S/FH.K/h1H;/q;+1/p-1
InChI key
NROKBHXJSPEDAR-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Quantity
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service