Skip to Content
Merck
All Photos(1)

Key Documents

52448

Supelco

Ranitidine S-oxide

pharmaceutical impurity standard, ≥95.0% (HPLC)

Synonym(s):

Ranitidine Related Compound C (USP), Ranitidine impurity C (PhEur), N-{2-{{{5-[(Dimethylamino)methyl]furan-2-yl}methyl}sulfinyl}ethyl}-N′-methyl-2-nitroethene-1,1-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22N4O4S
CAS Number:
Molecular Weight:
330.40
Beilstein:
8395257
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10.0% related substances
≤5.0% solvents
≤5.0% water

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN\C(NCCS(=O)Cc1ccc(CN(C)C)o1)=C/[N+]([O-])=O

InChI

1S/C13H22N4O4S/c1-14-13(9-17(18)19)15-6-7-22(20)10-12-5-4-11(21-12)8-16(2)3/h4-5,9,14-15H,6-8,10H2,1-3H3/b13-9+

InChI key

SKHXRNHSZTXSLP-UKTHLTGXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J H Kang et al.
Pharmacogenetics, 10(1), 67-78 (2000-03-30)
A non-invasive urine analysis method to determine the in-vivo flavin-containing mono-oxygenase (FMO) activity catalysing N-oxidation of ranitidine (RA) was developed and used to phenotype a Korean population. FMO activity was assessed by the molar concentration ratio of RA and RANO
P Viñas et al.
Journal of chromatography. B, Biomedical sciences and applications, 693(2), 443-449 (1997-06-06)
Ranitidine and its main metabolites, ranitidine N-oxide and ranitidine S-oxide, were determined in plasma and urine after separation using reversed-phase liquid chromatography. The mobile phase consisted of an initial isocratic step with 7:93 (v/v) acetonitrile-7.5 mM phosphate buffer (pH 6)
M H Davies et al.
Journal of hepatology, 22(5), 551-560 (1995-05-01)
We have previously reported an association of impaired S-oxidation with primary biliary cirrhosis. In order to confirm and further define this relationship, we retested S-oxidation capacity via three metabolic pathways and sulphation capacity via a fourth pathway. Metabolism of S-carboxymethyl-L-cysteine
M S Lant et al.
Journal of chromatography, 323(1), 143-152 (1985-04-17)
Reversed-phase high-performance liquid chromatography systems for the separation of ranitidine and its metabolites ranitidine-N-oxide, ranitidine-S-oxide, and desmethylranitidine have been developed for use in high-performance liquid chromatography-mass spectrometry. A direct liquid introduction-high-performance liquid chromatography-mass spectrometry system to analyse qualitatively and quantitatively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service