Skip to Content
Merck
All Photos(1)

Key Documents

50133

Supelco

Poly(vinyl chloride) carboxylated

Selectophore, 10% carboxyl basis

Synonym(s):

Poly(vinyl chloride-co-acrylic acid)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H4O2 · C2H3Cl)x
CAS Number:
MDL number:
UNSPSC Code:
26111700
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

mol wt

Mw ~175,000

concentration

10% (carboxyl)

SMILES string

ClC=C

InChI

1S/C3H4O2.C2H3Cl/c1-2-3(4)5;1-2-3/h2H,1H2,(H,4,5);2H,1H2

InChI key

SQNNHEYXAJPPKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IIIrd International Symposium on Neural Transplantation. Cambridge, 6-11 August, 1989. Proceedings.
Progress in brain research, 82, 1-734 (1990-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service