Skip to Content
Merck
All Photos(1)

Key Documents

45455

Supelco

Diphenamid

PESTANAL®, analytical standard

Synonym(s):

N,N-Dimethyl-2,2-diphenylacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2CHCON(CH3)2
CAS Number:
Molecular Weight:
239.31
Beilstein:
2052009
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

132-136 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

CN(C)C(=O)C(c1ccccc1)c2ccccc2

InChI

1S/C16H17NO/c1-17(2)16(18)15(13-9-5-3-6-10-13)14-11-7-4-8-12-14/h3-12,15H,1-2H3

InChI key

QAHFOPIILNICLA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Aharonson et al.
Archives of insect biochemistry and physiology, 22(3-4), 451-466 (1993-01-01)
Recent studies have demonstrated that interaction between various agrochemicals and soil microorganisms may either slow down or enhance processes of degradation. Soil disinfestation is employed for the control of soil-borne pathogens and weeds. Soil application of such broad-spectrum biocides, as
Hai-chao Liang et al.
Chemosphere, 80(4), 366-374 (2010-05-25)
In this work, the homogeneous and heterogeneous degradations of diphenamid (DPA) in aqueous solution were conducted by direct photolysis with UVC (254nm) and by photocatalysis with TiO(2)/UVA (350nm), and the experimental results were compared. It was found that the homogeneous
[Data on establishing the maximum permissible concentration of difenamid in the air of a work area].
I L Medved'
Gigiena i sanitariia, (5)(5), 77-78 (1983-05-01)
R D Verschoyle et al.
Fundamental and applied toxicology : official journal of the Society of Toxicology, 18(1), 79-88 (1992-01-01)
The insect repellent DEET and the structurally related herbicide diphenamid both cause ataxia associated with a spongiform myelinopathy largely confined to the cerebellar roof nuclei. This local myelinopathy was accompanied by the formation of neuronal cytoplasmic clefts and was produced
Hongxia Liu et al.
Annali di chimica, 95(5), 369-374 (2006-02-16)
Diphenamide, napropamide and metolachlor (FIG. 1) are selective, pre-emergence arylamide herbicides used to control the growth of annual grasses and broadleaf weeds in a variety of fields, e.g. fruit trees, nuts, corns, green crops, etc. They possess high activity and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service