Skip to Content
Merck
All Photos(2)

Key Documents

45427

Supelco

Desmetryn

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15N5S
CAS Number:
Molecular Weight:
213.30
Beilstein:
612017
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CNc1nc(NC(C)C)nc(SC)n1

InChI

1S/C8H15N5S/c1-5(2)10-7-11-6(9-3)12-8(13-7)14-4/h5H,1-4H3,(H2,9,10,11,12,13)

InChI key

HCRWJJJUKUVORR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>212.0 °F

Flash Point(C)

> 100 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Núria Borràs et al.
Chemosphere, 85(7), 1167-1175 (2011-10-15)
The mineralization of acidic aqueous solutions of the herbicide desmetryne has been studied by electrochemical advanced oxidation processes (EAOPs) such as anodic oxidation with electrogenerated H(2)O(2) (AO-H(2)O(2)), electro-Fenton (EF) and photoelectro-Fenton (PEF) with UVA light. Electrolyses were conducted in an
Michael P Alexander et al.
Journal of cognitive neuroscience, 21(7), 1422-1434 (2008-08-30)
List-learning tasks are frequently used to provide measures of "executive functions" that are believed necessary for successful memory performance. Small sample sizes, confounding anomia, and incomplete representation of all frontal regions have prevented consistent demonstration of distinct regional frontal effects
M N Poglazova et al.
Mikrobiologiia, 55(5), 825-830 (1986-09-01)
The work was aimed at studying how the herbicide semerone affected the ultrastructure of two soil microorganisms, Mycobacterium rubrum and Streptomyces bacillaris. Depending on its concentration, the herbicide inhibited growth processes so that biomass yield decreased, cell division was interfered
[Effect of the herbicide, semeron, on the food value and safety indices of cabbage].
M M Dubenetskaia et al.
Gigiena i sanitariia, (7)(7), 76-76 (1980-07-01)
[Hygienic establishment of the maximum permissible concentration of semeron in the soil].
N F Motuzinskiĭ et al.
Gigiena i sanitariia, (3)(3), 77-78 (1988-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service