Skip to Content
Merck
All Photos(1)

Key Documents

40927

Supelco

Neohesperidin

analytical standard

Synonym(s):

(S)-4′-Methoxy-3′,5,7-trihydroxyflavanone-7-[2-O-(α-L-rhamnopyranosyl)-β-D-glucopyranoside], Hesperetin 7-O-neohesperidoside, NSC 31048

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H34O15
CAS Number:
Molecular Weight:
610.56
Beilstein:
74945
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water (Karl Fischer)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(cc1O)[C@@H]2CC(=O)c3c(O)cc(O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O[C@@H]5O[C@@H](C)[C@H](O)[C@@H](O)[C@H]5O)cc3O2

InChI

1S/C28H34O15/c1-10-21(33)23(35)25(37)27(39-10)43-26-24(36)22(34)19(9-29)42-28(26)40-12-6-14(31)20-15(32)8-17(41-18(20)7-12)11-3-4-16(38-2)13(30)5-11/h3-7,10,17,19,21-31,33-37H,8-9H2,1-2H3/t10-,17-,19+,21-,22+,23+,24-,25+,26+,27-,28+/m0/s1

InChI key

ARGKVCXINMKCAZ-UZRWAPQLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Flavanone glycoside with neuroprotective and antioxidant properties. Does not inhibit oral carcinogenesis in a rat model unlike other citrus flavanones, but does exert a protective effect against gastritis and gastric lesions.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Su-Chen Ho et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 71, 176-182 (2014-06-24)
Inhibiting microglial activation-mediated neuroinflammation has become a convincing target for the development of functional foods to treat neurodegenerative diseases. Tangerine peel (Citri reticulatae pericarpium) has potent anti-inflammatory capacity; however, its anti-neuroinflammatory capacity and the corresponding active compounds remain unclear. To
Feng-Quan Tan et al.
BMC plant biology, 15, 89-89 (2015-04-08)
Polyploidy has often been considered to confer plants a better adaptation to environmental stresses. Tetraploid citrus rootstocks are expected to have stronger stress tolerance than diploid. Plenty of doubled diploid citrus plants were exploited from diploid species for citrus rootstock
Chung-Hsi Chou et al.
Food chemistry, 168, 63-69 (2014-08-31)
Black vinegar (BV) contains abundant essential and hydrophobic amino acids, and polyphenolic contents, especially catechin and chlorogenic acid via chemical analyses. K and Mg are the major minerals in BV, and Ca, Fe, Mn, and Se are also measured. After
Wanpeng Xi et al.
Food chemistry, 161, 230-238 (2014-05-20)
China is one of the most important diversity centres of the genus Citrus L. and is particularly rich in pummelo germplasm. In this study, the flavonoids in the peels and pulps of 28 Chinese local pummelos and four grapefruits were
Sylwia Magiera et al.
Journal of pharmaceutical and biomedical analysis, 102, 468-475 (2014-12-03)
A simple and rapid ultra-high performance liquid chromatographic (UHPLC) method coupled with an ultraviolet detector (UV) has been developed and validated for the separation and determination of 14 major flavonoids ((±)-catechin, (-)-epicatechin, glycitin, (-)-epicatechin gallate, rutin, quercitrin, hesperidine, neohesperidine, daidzein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service