Skip to Content
Merck
All Photos(1)

Key Documents

34980

Sigma-Aldrich

Dibutyltin(IV) oxide

purum

Synonym(s):

Dibutyloxotin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)2SnO
CAS Number:
Molecular Weight:
248.94
Beilstein:
4126243
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

autoignition temp.

534 °F

mp

≥300 °C (lit.)

SMILES string

CCCC[Sn](=O)CCCC

InChI

1S/2C4H9.O.Sn/c2*1-3-4-2;;/h2*1,3-4H2,2H3;;

InChI key

JGFBRKRYDCGYKD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dibutyltin(IV) oxide (Bu2SnO) can be used as a reagent to prepare:
  • Substituted oxazoles via reaction of dibutyltin diacylates with 1-substituted acetylenes and trimethylsilyl azide.
  • Triazolyl-substituted benzyloxyacetohydroxamic acids as potential LpxC inhibitors.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Muta. 2 - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 - STOT SE 1

Target Organs

thymus

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qinghui Wang et al.
Carbohydrate research, 342(17), 2657-2663 (2007-09-26)
Regioselective formation of 6-O-acylsucroses and 6,3'-di-O-acylsucroses in one pot with good yields was achieved for the first time by a typical acylation method of sucrose via its dibutylstannylene acetal. Pure monoesters at OH-6 and diesters at OH-6,3' obtained by these
[Experimental toxicity of indium formate and a mixture of indium formate-dibutyltin oxide (90-10) in the rat].
J P Anger et al.
Journal de toxicologie clinique et experimentale, 8(6), 401-418 (1988-11-01)
Shao-Min Wang et al.
Steroids, 72(1), 26-30 (2006-12-30)
Dibutyltin oxide (DBTO) was first utilized for the deacetylation of steroid and diterpene esters. The results showed the deprotection of acetylated steroids and diterpenes separately with moderate catalysis dibutyltin oxide in methanol selectively removed part acetyl groups of these substrates
K Kiyoshima et al.
Chemical & pharmaceutical bulletin, 37(4), 861-865 (1989-04-01)
4''-O-Acyl-, 4''-O-alkyl- and 4''-deoxy-tylosin derivatives were synthesized using 2'-O-acetyl-3'',4''-O-(dibutyl-stannio)tylosin as a synthetic intermediate. The in vitro biological evaluation showed that the new derivatives were active against macrolide-resistant clinical isolates of bacteria and mycoplasmas, and that they were resistant to hepatic
Shao-Min Wang et al.
Carbohydrate research, 346(2), 203-209 (2010-12-28)
The reaction process for the selective deprotection of acetylated glucosides by dibutyltin oxide in methanol is investigated by using methyl 2,3,4,6-tetra-O-acetyl-α-d-glucopyranoside as a model substrate with ESIMS and NMR techniques. According to the results, it is inferred that at first

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service