Skip to Content
Merck
All Photos(2)

Key Documents

34027

Supelco

Fluazifop-methyl

PESTANAL®, analytical standard

Synonym(s):

Methyl 2-[4-(5-trifluoromethyl-2-pyridyloxy)phenoxy]propionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14F3NO4
CAS Number:
Molecular Weight:
341.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

75-80 °C

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)C(C)Oc1ccc(Oc2ccc(cn2)C(F)(F)F)cc1

InChI

1S/C16H14F3NO4/c1-10(15(21)22-2)23-12-4-6-13(7-5-12)24-14-8-3-11(9-20-14)16(17,18)19/h3-10H,1-2H3

InChI key

IAUMNRCGDHLAMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fluazifop-methyl belongs to the aryloxyphenoxyacid class of herbicides.

Application

Fluazifop-methyl may be used as an analytical reference standard for the determination of the analyte in soybeans, soil and potatoes using capillary gas-liquid chromatography with a nitrogen-specific detector.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of herbicide residues in cereals, fruits and vegetables.
Tadeo LJ, et al.
Journal of Chromatography A, 882(1-2), 175-191 (2000)
Gas chromatographic analysis of fluazifop-butyl (Fusilade) in potatoes, soybeans, and soil.
Clegg SB.
Journal of Agricultural and Food Chemistry, 35(2), 269-273 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service