33559
4-tert-Octylphenol-3,5-d2 solution
1 μg/mL in acetone, analytical standard
Synonym(s):
4-(1,1,3,3-Tetramethylbutyl)phenol-3,5-d2, 4-tert-OP-D2
About This Item
grade
analytical standard
shelf life
limited shelf life, expiry date on the label
concentration
1 μg/mL in acetone
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
application(s)
environmental
format
single component solution
mass shift
M+2
storage temp.
−20°C
SMILES string
[2H]c1cc(O)cc([2H])c1C(C)(C)CC(C)(C)C
InChI
1S/C14H22O/c1-13(2,3)10-14(4,5)11-6-8-12(15)9-7-11/h6-9,15H,10H2,1-5H3/i6D,7D
InChI key
ISAVYTVYFVQUDY-QFIQSOQBSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
1.4 °F - closed cup
Flash Point(C)
-17.0 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Alkylphenols are starting materials for the synthesis of alkylphenol ethoxylates, which are widely used as non ionic tensides, dispersive agents in paper and leather manufacturing, emulsifiers for pesticide formulations and as auxiliary agents for drilling and flotation.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service