Skip to Content
Merck
All Photos(1)

Key Documents

18543

Supelco

Acetoxyvalerenic acid

analytical standard

Synonym(s):

Acetylvalerenolic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H24O4
CAS Number:
Molecular Weight:
292.37
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤5.0% water

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

C[C@@H]1CC[C@@H](\C=C(/C)C(O)=O)C2=C(C)C[C@@H](OC(C)=O)[C@H]12

InChI

1S/C17H24O4/c1-9-5-6-13(7-11(3)17(19)20)15-10(2)8-14(16(9)15)21-12(4)18/h7,9,13-14,16H,5-6,8H2,1-4H3,(H,19,20)/b11-7+/t9-,13+,14-,16+/m1/s1

InChI key

VBBXZFLAYWAXSK-HYJCDKNOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetoxyvalerenic acid is a derivative of valerenic acid. It is a component of essential oil of valerian. It has hypnotic effect.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: valeriana

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Gail D Anderson et al.
Phytotherapy research : PTR, 24(10), 1442-1446 (2010-09-30)
Insomnia is a commonly reported clinical problem with as many as 50% of older adults reporting difficulty in falling and/or remaining asleep. Valerian (Valeriana officinalis) is a commonly used herb that has been advocated for promoting sleep. Valerenic acid is
Juliane Hintersteiner et al.
European journal of pharmacology, 735, 123-131 (2014-04-01)
Valerenic acid (VA) is a β2/3 subunit-specific modulator of γ-aminobutyric acid (GABA) type A (GABAA) receptors inducing anxiolysis. Here we analyze if VA-esters can serve as prodrugs and if different ester structures have different in vitro/in vivo effects. Modulation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service