Skip to Content
Merck
All Photos(1)

Key Documents

134210

Gardner Colour Reference Standard

ASTM D 1544, D 6166 (ASTM colour 5)

Synonym(s):

Color Reference Standard Gardner 5, Colour Reference Standard Gardner 5, Gardner Color Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

according to ASTM® D 1544
according to ASTM® D 6166

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

This product is a certified reference material (CRM), traceable to international standards BS EN ISO / IEC 17025 and ISO 17034 as per UKAS accreditation.

Please note that the value stated on this page is nominal. All certified values for the current lots of all Gardner color reference standards can be found on the Paragon Scientific webpage.

Application

Colour Reference Standard Gardner 5 is used to calibrate color measuring instruments and validate test results.
It finds use in process industries that employ colorimeters to analyze the colour control of products such as oils, fuels, chemicals, pharmaceuticals, paints and coatings.

Features and Benefits

  • Manufactured and tested according to ISO/IEC 17025 and ISO 17034 under UKAS accreditation
  • Colour values within ASTM D 1544/D 6166 colour scale
  • Supplied with a certificate of calibration and MSDS
  • Stored in a 500-mL tamper-evident bottles
  • Shelf-life of 18-months

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

replaced by

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service