Skip to Content
Merck
All Photos(1)

Key Documents

134170

Pt-Co/Hazen/APHA Colour Reference Standard

ASTM D 1209 (ASTM colour 30)

Synonym(s):

Colour Reference Standard Platinum Cobalt 30, Pt-Co/Hazen/APHA Color Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Paragon Scientific Ltd

application(s)

cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical

format

mixture

Looking for similar products? Visit Product Comparison Guide

General description

This product is an analytical standard, complying ISO 9001 Quality system. It's certified using a master instrument and reference standards by measuring spectral transmittance as a key property of calibration.

Please note that the value stated on this page is nominal. All certified values for the current lots of all Pt-Co/Hazen/APHA Colour Reference Standards can be found on the Paragon Scientific webpage.

Application

Pt-Co/Hazen/APHA (ASTM colour 30) colour reference standard is suitable for use in standard test methods for colour of liquids of the platinum-cobalt scale.

It may be used as a colour standard for visual colour measurement of water samples according to ASTM D 1209 method.

Features and Benefits

  • Qualified under ISO 9001 Quality management system
  • Accompanied with a certificate of analysis and MSDS
  • Full traceability to international standards with maximum data accuracy
  • Colour values conforming to ASTM D 1209 colour scale
  • Supplied in a 500-mL pack size with tamper-evident packaging
  • 12-months of shelf life

replaced by

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service