Skip to Content
Merck
All Photos(1)

Key Documents

CBL1358

Sigma-Aldrich

Anti-Endoglin Antibody, Extracellular, clone MJ7/18

clone MJ7/18, Chemicon®, from rat

Synonym(s):

CD105, Ancillary TGF-beta Receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
MJ7/18, monoclonal
application:
FACS
IHC
IP
WB
species reactivity:
mouse
technique(s):
flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
citations:
2

biological source

rat

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MJ7/18, monoclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

suitability

not suitable for immunohistochemistry (Paraffin)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Eng(13805)

General description

Endoglin is the regulatory component of the TGF-beta receptor complex. CD105 is a homodimer of 90 kDa subunits and is predominantly expressed on vascular endothelial cells. It is also found on pre-erythroblasts, macrophages and lymphoid and myeloid leukemic cells High levels of mouse endoglin mRNA have been detected in ovary, uterus, and NCTC-2071 fibroblasts, and to a lesser extent, in heart and muscle. In addition, stromal cells in the connective tissue of various organs are also positive for endoglin expression. CD105 has been reported to have functions in adhesion and embryonic angiogenesis, and that MJ7/18 is a pan endothelial marker (with minimal reactivity with early hematopoetic cells). Both mouse and human CD105 exhibit >70% sequence similarity in their cytoplasmic domains with type III transforming growth factor β (TGFβ) receptor.

Specificity

Specifically recognizes mouse CD105, known as endoglin.

Immunogen

Inflamed mouse skin (Ge, AZ et al, 1994).

Application

Flow Cytometry: 1μg/ 10(e6) cells in 100 μL total volume.

Immunohistochemical staining of fresh frozen or acetone fixed tissue specimens.1:10-1:50. Not recommended for formalin fixed paraffin tissues.

Immunoprecipitation: Use rabbit anti-rat secondary antibody followed by protein A, or anti-rat beads for recovery.

Western Blotting: 1:500-1:1000. Recognizes a 90 kDa band under reducing conditions. Membrane preparations are recommended. Mouse vascular endothelial cells recommended as a positive control.

Optimal working dilutions must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins
This Anti-Endoglin Antibody, Extracellular, clone MJ7/18 is validated for use in FC, IP, WB, IH for the detection of Endoglin.

Physical form

Format: Purified
Protein A purified immunoglobulin presented as a liquid in 100 mM borate buffered saline, containing no preservatives.

Storage and Stability

Maintain at 2-8°C for up to 12 months from date of receipt.

Analysis Note

Control
POSITIVE CONTROL: mouse vascular endothelial cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Z Ge et al.
Gene, 138(1-2), 201-206 (1994-01-28)
The rat monoclonal antibody, MJ7/18, which reacts selectively with the endothelium of blood vessels in mouse was used to screen a cDNA library derived from a transformed mouse brain endothelial cell line. The sequence of a cDNA encoding the cell
Nanoparticle-mediated drug delivery to tumor vasculature suppresses metastasis.
Murphy, EA; Majeti, BK; Barnes, LA; Makale, M; Weis, SM; Lutu-Fuga, K; Wrasidlo, W; Cheresh, DA
Proceedings of the National Academy of Sciences of the USA null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service