Skip to Content
Merck
All Photos(1)

Key Documents

8.02815

Sigma-Aldrich

Cyanuric chloride

for synthesis

Synonym(s):

Cyanuric chloride, 2,4,6-Trichloro-1,3,5-triazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3Cl3N3
CAS Number:
Molecular Weight:
184.41
MDL number:
UNSPSC Code:
12352101
EC Index Number:
203-614-9

vapor pressure

0.01 hPa ( 20 °C)

Quality Level

form

powder

autoignition temp.

>650 °C

potency

315 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

reaction suitability

reagent type: oxidant

bp

193-198 °C/1013 hPa

mp

144-146 °C

transition temp

flash point >190 °C

density

1.92 g/cm3 at 20 °C

bulk density

600‑900 kg/m3

storage temp.

2-30°C

InChI

1S/C3Cl3N3/c4-1-7-2(5)9-3(6)8-1

InChI key

MGNCLNQXLYJVJD-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 99.0 %
Melting range (lower value): ≥ 144 °C
Melting range (upper value): ≤ 147 °C
Identity (IR): passes test

Pictograms

CorrosionSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1A - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

392.0 °F - closed cup

Flash Point(C)

> 200 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service