6570
L-Tyrosine, Free Base
Non-essential amino acid for human development. Precursor for the synthesis of thyroid hormones and select neurotransmitters, such as dopamine and norepinephrine.
Synonym(s):
L-Tyrosine, Free Base, Tyr
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H11NO3
CAS Number:
Molecular Weight:
181.19
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.54
Recommended Products
Quality Level
Assay
≥98% (titration)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
white
cation traces
heavy metals: ≤30 ppm
shipped in
ambient
storage temp.
15-25°C
InChI
1S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11H,5,10H2,(H,12,13)
InChI key
OUYCCCASQSFEME-UHFFFAOYSA-N
General description
Non-essential amino acid for human development. Precursor for the synthesis of thyroid hormones and select neurotransmitters, such as dopamine and norepinephrine.
Warning
Toxicity: Irritant (B)
Preparation Note
Solubility in H₂O is higher at alkaline pH.
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service