516090-M
Penciclovir
A carba analog of Ganciclovir. Active against several herpes viruses. Induces apoptosis effectively without exerting much genotoxicity.
Synonym(s):
Penciclovir, 2-Amino-1,9-dihydro-9-[4-hydroxy-3-(hydroxymethyl)butyl]-6H-purin-6-one
About This Item
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
color
white
solubility
water: 1 mg/mL
aqueous buffer, pH 2.0: 10 mg/mL
storage temp.
−20°C
InChI
1S/C10H15N5O3/c11-10-13-8-7(9(18)14-10)12-5-15(8)2-1-6(3-16)4-17/h5-6,16-17H,1-4H2,(H3,11,13,14,18)
InChI key
JNTOCHDNEULJHD-UHFFFAOYSA-N
General description
Warning
Reconstitution
Other Notes
Amjad, M., et al. 2001. Microbiol. Immunol.45, 233.
Thust, R., et al. 2000. Cancer Gene Ther.7, 107.
Thust, R., et al. 1996. Antiviral Res.31, 105.
Shaw, T., et al. 1994. Antimicrob. Agents Chemother.38, 719.
Harnden, M.R. 1989. J. Med. Chem.32, 1738.
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service