07-753
Anti-RIAP-3 (XIAP) Antibody
Upstate®, from rabbit
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Recommended Products
biological source
rabbit
antibody form
purified antibody
antibody product type
primary antibodies
clone
polyclonal
species reactivity
rat, human
manufacturer/tradename
Upstate®
technique(s)
western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
human ... XIAP(331)
Specificity
RIAP-3 / XIAP
Immunogen
GST fusion protein corresponding to full-length rat inhibitor of apoptosis 3 (RIAP-3), also know as X-linked inhibitor of apoptosis (XIAP)
Application
Anti-RIAP-3 (XIAP) Antibody detects level of RIAP-3 (XIAP) & has been published & validated for use in WB.
Research Category
Apoptosis & Cancer
Apoptosis & Cancer
Research Sub Category
BCL2 & Inhibition
BCL2 & Inhibition
Quality
Routinely evaluated by immunoblot on RIPA lysates from Jurkat cells
Target description
55 kDa; an additional protein was also detected at Mr 120 kDa
Physical form
Format: Purified
Protein A purified
Storage and Stability
2 years at -20°C from date of shipment
Analysis Note
Control
Positive Antigen Control: Catalog #12-303, Jurkat cell lysate.
Positive Antigen Control: Catalog #12-303, Jurkat cell lysate.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
cIAP-1, but not XIAP, is cleaved by caspases during the apoptosis induced by TGF-beta in fetal rat hepatocytes.
Herrera, Blanca, et al.
Febs Letters, 520, 93-96 (2002)
Guy S Salvesen et al.
Nature reviews. Molecular cell biology, 3(6), 401-410 (2002-06-04)
The 'inhibitor of apoptosis' (IAP) gene family, which was discovered less than a decade ago, encodes a group of structurally related proteins that, in addition to their ability to suppress apoptotic cell death, are involved in an increasing number of
Y Sonoda et al.
The Journal of biological chemistry, 275(21), 16309-16315 (2000-05-24)
Focal adhesion kinase (FAK) has an anti-apoptotic role in anchorage-dependent cells via an unknown mechanism. To elucidate the role of FAK in anti-apoptosis, we have established several FAK cDNA-transfected HL-60 cell lines and examined whether FAK-transfected cells have resistance to
Andreas Bergmann et al.
Current opinion in cell biology, 15(6), 717-724 (2003-12-04)
Inhibitor of apoptosis proteins (IAPs) are a conserved class of proteins that control apoptosis in both vertebrates and invertebrates. They exert their anti-apoptotic function through inhibition of caspases, the principal executioners of apoptotic cell death. Recent advances in vertebrates and
Robert J Tower et al.
eLife, 11 (2022-05-27)
De novo limb regeneration after amputation is restricted in mammals to the distal digit tip. Central to this regenerative process is the blastema, a heterogeneous population of lineage-restricted, dedifferentiated cells that ultimately orchestrates regeneration of the amputated bone and surrounding
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service