TEM-CF200CU50
Carbon Film Supported Copper Grid
Standard Thickness, size 200 mesh, box of 50
Synonym(s):
Carbon Film 200 Mesh, Copper
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41111761
Recommended Products
material
carbon film
copper grid
packaging
box of 50
film size
5-6 nm
size
200 mesh
Looking for similar products? Visit Product Comparison Guide
General description
TEM grids supported by carbon film only. A 5-6 nm thin film of pure carbon deposited on one side of the standard square mesh grid.
Application
- Chemical vapor deposition growth of linked carbon monolayers with acetylenic scaffoldings on silver foil: This study discusses the use of carbon films supported on copper grids to analyze the structural and electrical properties of linked carbon monolayers grown via chemical vapor deposition (CVD). The analysis included SEM and TEM imaging, which demonstrated the effective suspension of the film on TEM copper grids (R Liu et al., 2017).
- Surface monocrystallization of copper foil for fast growth of large single-crystal graphene under free molecular flow: This research discusses the preparation and use of carbon film supported copper grids for the synthesis of large single-crystal graphene, highlighting the technique′s potential for creating high-quality graphene sheets (H Wang et al., 2016).
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service