900703
Methoxy poly(ethylene glycol)-b-poly(L-lactide)
2k-2k
Synonym(s):
mPEG-b-PLA, mPEG-PLA, mPEG-PLLA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
HO[CH(CH3)COO]m[CH2CH2O]nCH3
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
form
powder
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
Biocompatible, amphiphilic block copolymer composed of a hydrophilic PEG block and a hydrophobic poly(L-lactide) (PLLA) block. These materials have been used in control release and nanoparticle formulation for drug encapsulation and delivery applications. Well-defined materials with varying properties can be prepared by controlling the relative length of each polymer block. Hydroxyl termination allows for facile further chemical modification of these materials.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Zhigang Xie et al.
Journal of biomedical materials research. Part A, 88(1), 238-245 (2008-02-21)
Docetaxel (DX) is one of the most effective antineoplastic drugs. Its current clinical administration is limited because of its hydrophobicity and serious side effects. A polymer/DX conjugate is designed and successfully prepared to solve these problems. It is monomethoxy-poly(ethylene glycol)-block-poly(L-lactide)/DX
Injectable biodegradable polymeric system for preserving the active form and delayed-release of camptothecin anticancer drugs.
Mert O, et al.
Royal Society of Chemistry Advances, 2(1), 176-185 (2012)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service