808814
Molybdenum telluride
Crystal, 99.995%
Synonym(s):
Molybdenum (IV) telluride, Molybdenum ditelluride
About This Item
Recommended Products
Quality Level
Assay
99.995%
form
crystals
L × W × H
0.5 cm × 0.5 cm × 0.2-0.3 mm, typical size, some variation in dimensions may occur with different batches
SMILES string
[Te]=[Mo]=[Te]
InChI
1S/Mo.2Te
InChI key
HITXEXPSQXNMAN-UHFFFAOYSA-N
Application
Preparation Note
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Acute Tox. 4 Inhalation
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Advances in scalable synthesis and processing of two-dimensional materials
For several decades, the need for an environmentally sustainable and commercially viable source of energy has driven extensive research aimed at achieving high efficiency power generation systems that can be manufactured at low cost.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service