Skip to Content
Merck
All Photos(3)

Key Documents

796492

Sigma-Aldrich

Silver nanoplates

resonant wavelength, 750 nm, 0.02 mg/mL (in water with 5 mM sodium borate buffer), PVP functionalized

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ag
Molecular Weight:
107.87
UNSPSC Code:
12352300
NACRES:
NA.23

form

nanoplates

concentration

0.02 mg/mL (in water with 5 mM sodium borate buffer)

resonant wavelength

750 nm

functional group

PVP

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Store products away from light at 4-6°C. Short periods at room temperature are acceptable however, lower temperature storage prolongs the shelf life of the product. DO NOT FREEZE.

Application

Silver nanomaterials have unique physical, chemical and optical properties that are currently being leveraged for a wide variety of biological applications.,,,,,,,, A resurgence of interest in the utility of silver as a broad based antimicrobial agent has led to the development of hundreds of products that incorporate silver nanoparticles to prevent bacterial growth on surfaces.,,, Additionally, silver nanoparticles have an optical color that is a function of the silver nanoparticle′s size and shape.The strong coupling of silver nanoparticles to specific wavelengths of incident light can be utilized to develop ultrabright reporter molecules,,,, highly efficient thermal absorbers, and nanoscale “antennas” that can amplify the strength of the local electromagnetic field.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surface roughness and the enhanced intensity of Raman scattering by molecules adsorbed on metals
Moskovits M, et al.
J. Chem. Phys., 69, 4159-4159 (1978)
Characterization of the size, shape, and state of dispersion of nanoparticles for toxicological studies
Powers KW, et al.
Nanotoxicology, 1(1), 42-51 (2007)
A review of the antibacterial effects of silver nanomaterials and potential implications for human health and the environment.
Jones C, et al.
Journal of Nanoparticle Research, 12(5), 1531-1551 (2010)
C Carlson et al.
The journal of physical chemistry. B, 112(43), 13608-13619 (2008-10-04)
The rapid advancement of nanotechnology has created a vast array of engineered nanomaterials (ENMs) which have unique physical (size, shape, crystallinity, surface charge) and chemical (surface coating, elemental composition and solubility) attributes. These physicochemical properties of ENMs can produce chemical
Single Molecule Detection Using Surface-Enhanced Raman Scattering (SERS)
Kneipp K, et al.
Physical Review Letters, 78, 1667-1667 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service