Skip to Content
Merck
All Photos(1)

Key Documents

769312

Sigma-Aldrich

Chromium(III) chloride hexahydrate

≥96%

Synonym(s):

Chromium trichloride hexahydrate, Hexaaquachromium(III) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CrCl3 · 6H2O
CAS Number:
Molecular Weight:
266.45
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥96%
96.0-104.0% (RT)
form:
solid

Assay

≥96%
96.0-104.0% (RT)

form

solid

pH

2.0-3.0

density

1.76 g/mL at 25 °C (lit.)

anion traces

sulfate (SO42-): ≤0.05%

cation traces

Cu: ≤0.001%
Fe: ≤0.03%
Pb: ≤0.005%

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].Cl[Cr](Cl)Cl

InChI

1S/3ClH.Cr.6H2O/h3*1H;;6*1H2/q;;;+3;;;;;;/p-3

InChI key

LJAOOBNHPFKCDR-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Salts of chromium are used as precursor in synthesis of chromium based catalysts and nanoparticles. Chromium salts also find applications in electrodeposition of corrosion and wear resistant coatings.

Analysis Note

Substances not precipitated by ammonia (as sulphate) ≤ 0.2 %

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Met. Corr. 1 - Skin Sens. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Facile method to attach transition metal ions to the surface of anatase TiO(2) nanorods.
Balasanthiran, C., et al.
Chemical Communications (Cambridge, England), 50(43), 5721-5724 (2014)
Synthesis of Bimetallic Ni-Cr Nano-Oxides as Catalysts for Methanol Oxidation in NaOH Solution
Gu, Y., et al.
Journal of Nanoscience and Nanotechnology, 15(5), 3743-3749 (2015)
Speciation, physical and electrolytic properties of eutectic mixtures based on CrCl3?6H2O and urea
Abbott, AP., et al.
Physical Chemistry Chemical Physics, 16, 9047-9055 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service