Skip to Content
Merck
All Photos(1)

Key Documents

752193

Sigma-Aldrich

1,3,5-Tris(N-carbazolyl)benzene

97%

Synonym(s):

1,3,5-Tris(carbazol-9-yl)benzene, TCB, tCP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H27N3
CAS Number:
Molecular Weight:
573.68
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

325-330 °C

Orbital energy

HOMO 5.8 eV 
LUMO 2.3 eV 

SMILES string

c1ccc2c(c1)n(-c3cc(cc(c3)-n4c5ccccc5c6ccccc46)-n7c8ccccc8c9ccccc79)c%10ccccc2%10

InChI

1S/C42H27N3/c1-7-19-37-31(13-1)32-14-2-8-20-38(32)43(37)28-25-29(44-39-21-9-3-15-33(39)34-16-4-10-22-40(34)44)27-30(26-28)45-41-23-11-5-17-35(41)36-18-6-12-24-42(36)45/h1-27H

InChI key

DVNOWTJCOPZGQA-UHFFFAOYSA-N

Application

This material is an advanced host material for phosphorescent guest-host systems; such as used in Organic Light Emitting Diode devices.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pascal Duenk et al.
Genetics, selection, evolution : GSE, 51(1), 38-38 (2019-07-10)
Pig and poultry breeding programs aim at improving crossbred (CB) performance. Selection response may be suboptimal if only purebred (PB) performance is used to compute genomic estimated breeding values (GEBV) because the genetic correlation between PB and CB performance ([Formula:
Thoms, T.; Okada, S.; Chen, J. P.; Furugori, M.
Thin Solid Films, 436, 264-264 (2003)
Barbara Lapinska et al.
Molecules (Basel, Switzerland), 24(3) (2019-01-27)
This study investigated the effect of saliva contamination on chemical changes of ceramic surface as well as the influence of saliva cleaning methods on ceramic-resin bond strength. Saliva was used to contaminate leucite (LGC) and lithium disilicate (LDGC) glass ceramic
Min Sun et al.
Veterinary microbiology, 205, 6-13 (2017-06-18)
Porcine epidemic diarrhea virus (PEDV), is the etiological agent of porcine epidemic diarrhea virus (PED), causes high mortality with severe vomiting, diarrhea, and dehydration in swine farms. In this study, the PEDV strain 85-7 could be proliferated effectively in MARC-145
Yonggang Liu et al.
Journal of chromatography. A, 1522, 23-29 (2017-10-02)
A propylene-ethylene random copolymer was fractionated by preparative temperature rising elution fractionation (TREF). The structural heterogeneity of the bulk sample and its TREF fractions was studied by high temperature liquid chromatography with a solvent gradient elution from 1-decanol to 1,2,4-trichlorobenzene.

Articles

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service