Skip to Content
Merck
All Photos(1)

Key Documents

731048

Sigma-Aldrich

Poly(N-isopropylacrylamide), maleimide terminated

average Mn 2,000

Synonym(s):

P(NIPAM)n-NH-maleimide, Polyacrylamide, functionalized polyNIPAM, functionalized polyacrylamide, polyNIPAM

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11NO)n SCH2CH2NH(CO)CH2CH2CH2(C4H2NO2)
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

average Mn 2,000

transition temp

Tm 216-262 °C

Looking for similar products? Visit Product Comparison Guide

Application

Novel polymer for development of thermosensitive coated micro/nano materials including thermoresponsive polymeric drug delivery systems. The maleimide functional group can be used to conjugate a variety of biomolecules such as enzymes and DNA to the polymer chain.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chung, J. E.; Yokoyama, M.; Suzuki, K.; Aoyagi, T.; Sakurai, Y.; Okano, T.
Colloids and Surfaces, B: Biointerfaces, 9, 37-37 (1997)
Safak, M.; Alemdaroglu, F. E.; Li, Y.; Ergen, E.; Hernnann, A.
Advanced Materials, 19, 1499-1499 (2007)
Karina L Heredia et al.
Macromolecules, 42(7), 2360-2367 (2009-01-01)
In this report we describe a straightforward approach to synthesize polymers with end-groups that bind site-specifically to two different proteins. Telechelic biotin, maleimide poly(
Hua Wei et al.
Biomaterials, 28(1), 99-107 (2006-09-09)
A four-arm star block copolymer, comprised of a hydrophobic PMMA arm and an average of three hydrophilic poly(N-isopropylacrylamide) (PNIPAAm) arms were designed and synthesized from the molecular level. The amphiphilic star block copolymer is capable of self-assembling into micelles in
K Fujimoto et al.
Biomacromolecules, 1(4), 515-518 (2001-11-17)
We report a novel drug delivery system for apoptosis induction by a "smart" polymer vehicle possessing thermosensitivity and bioaffinity. The polymer chain was prepared by copolymerization of N-isopropylacrylamide and N-methacryloyloxysuccinimide. Cell-adhesive RGDS peptide was conjugated with the copolymer as a

Articles

Poly(N-isopropylacrylamide), or PNIPAM, is a stimuli-responsive polymer that responds to changes in pH and temperature and has a LCST around 32 C.

Tissue engineering has become a key therapeutic tool in the treatment of damaged or diseased organs and tissues, such as blood vessels and urinary bladders.

By altering the physicochemical properties, smart or intelligent drug delivery systems can be designed to deliver therapeutic molecules on-demand. Learn more about the application of stimuli-responsive materials in drug delivery.

Wide range of functional polymers for biomedical applications have been synthesized and structurally characterized. Several classes of polymers including biodegradable polymers, hydrophilic & amphiphilic polymers, and stimuli responsive polymers have been prepared using controlled and directed functionalization via "living" polymerization such as RAFT, ionic and ring opening polymerization. Selected polymers have been studied for their structure-properties relationship. "

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service