563994
Nafion™ perfluorinated membrane
Nafion™ 424, reinforced with poly(tetrafluoroethylene) fiber, thickness 0.013 in.
About This Item
Recommended Products
eq. wt.
1,100
manufacturer/tradename
Nafion™ 424
L × W
12 in. × 12 in.
thickness
0.013 in.
solubility
H2O: insoluble
InChI
1S/C7HF13O5S.C2F4/c8-1(9)2(10)24-5(15,16)3(11,4(12,13)14)25-6(17,18)7(19,20)26(21,22)23;3-1(4)2(5)6/h(H,21,22,23);
InChI key
FOYUGSIADQEOEK-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
· Keep pieces sealed and protected from general exposure will reduce the effect. Recommend long term storage in air-tight bags.
· Discoloration does not affect performance and can be removed by soaking in dilute nitric acid.
Other Notes
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Proton exchange membrane (PEM) fuel cells operate at relatively low temperatures and are composed of two electrodes and a conductive elecrolyte.
Advances in the electrochemical conversion of water to and from hydrogen and oxygen have principally been achieved through the development of new materials and by understanding the mechanisms of the degradation of proton exchange membrane fuel cells (PEMFC) during operation.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service