Skip to Content
Merck
All Photos(1)

Key Documents

55999

Sigma-Aldrich

8-Hydroxyoctanoic acid

98% (T)

Synonym(s):

8-Hydroxycaprylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16O3
CAS Number:
Molecular Weight:
160.21
Beilstein:
1237092
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98% (T)

form

solid

mp

59-63 °C

functional group

carboxylic acid
hydroxyl

SMILES string

OCCCCCCCC(O)=O

InChI

1S/C8H16O3/c9-7-5-3-1-2-4-6-8(10)11/h9H,1-7H2,(H,10,11)

InChI key

KDMSVYIHKLZKET-UHFFFAOYSA-N

General description

8-Hydroxyoctanoic acid is one of the constituents of ‘‘Royal jelly′′ acid found in herbhoneys and genuine honeys.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical composition and antimicrobial activity of Polish herbhoneys.
Isidorov VA, et al.
Food Chemistry, 171, 84-88 (2015)
Biosynthesis, modification, and biodegradation of bacterial medium-chain-length polyhydroxyalkanoates.
Kim DY, et al.
Journal of Microbiology, 87-97 (2007)
Bora Kim et al.
Clinical pharmacology and therapeutics, 103(5), 879-887 (2017-09-07)
Quantitative models of endogenous metabolites are useful in predicting CYP3A-mediated drug-drug interactions. This study aimed to identify novel predictive markers for the magnitude of CYP3A induction and inhibition in male and female subjects using an untargeted metabolomics approach. Here we
Nicholas D Lanz et al.
Biochemistry, 55(9), 1372-1383 (2016-02-04)
The prevalence of multiple and extensively drug-resistant strains of Mycobacterium tuberculosis (Mtb), the causative agent of tuberculosis, is on the rise, necessitating the identification of new targets to combat an organism that has infected one-third of the world's population, according
Mahek Sadiq et al.
Biosensors, 11(2) (2021-02-10)
Major advances in cancer control can be greatly aided by early diagnosis and effective treatment in its pre-invasive state. Lung cancer (small cell and non-small cell) is a leading cause of cancer-related deaths among both men and women around the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service