Skip to Content
Merck
All Photos(2)

Key Documents

541346

Sigma-Aldrich

Nafion perfluorinated membrane

Nafion 115, thickness 0.005 in.

Synonym(s):

nafion membrane, proton exchange membrane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C7HF13O5S . C2F4)x
CAS Number:
MDL number:
UNSPSC Code:
26111700
eCl@ss:
32110203
NACRES:
NA.23

eq. wt.

1,100

manufacturer/tradename

Nafion 115

L × W

12 in. × 12 in.

thickness

0.005 in.

solubility

H2O: insoluble

InChI

1S/C7HF13O5S.C2F4/c8-1(9)2(10)24-5(15,16)3(11,4(12,13)14)25-6(17,18)7(19,20)26(21,22)23;3-1(4)2(5)6/h(H,21,22,23);

InChI key

FOYUGSIADQEOEK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nafion membranes are fluorinated polymers functionalized with sulphonic acid moieties.

Application

Nafion is a perfluorinated ion-exchange membrane which has a wide variety of commercial uses. The acidic (hydrogen ion) form and derivatives are solid, superacid catalysts useful in a wide variety of synthetic applications.
Nafion perfluorinated membrane is a composite membrane, which is utilized as an ion-exchange membrane for the fabrication of fuel cells. It can also be used as an electrolytic separator for the preparation of small scale supercapacitors.

Packaging

Darkening/browning is common for pieces stored in a general room environment for an extended period of time (several months), due to organics pick-up. Verify product is kept sealed during storage.
· Keep pieces sealed and protected from general exposure will reduce the effect. Recommend long term storage in air-tight bags.
· Discoloration does not affect performance and can be removed by soaking in dilute nitric acid.
Packaged in heat-sealed foil bag in stay-flat mailers

Legal Information

Nafion is a trademark of The Chemours Company FC, LLC

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Performance improvement of Nafion based solid state electrochemical supercapacitor
Lufrano F and Staiti P
Electrochimica Acta, 49(16), 2683-2689 (2004)
Nafion perfluorinated membranes in fuel cells
Banerjee S and Curtin DE
Journal of Fluorine Chemistry, 125(8), 1211-1216 (2004)
Mustafa Bener et al.
Analytical chemistry, 82(10), 4252-4258 (2010-04-27)
A low-cost optical sensor using an immobilized chromogenic redox reagent was devised for measuring the total antioxidant level in a liquid sample without requiring sample pretreatment. The reagent, copper(II)-neocuproine (Cu(II)-Nc) complex, was immobilized onto a cation-exchanger film of Nafion, and
Min-Ho Park et al.
Advanced materials (Deerfield Beach, Fla.), 27(29), 4308-4314 (2015-06-24)
A novel flexible encapsulation method (Flex Lami-capsulation) is reported, which can be applied in the roll-to-roll process for mass production of organic electronic devices. Flex Lami-capsulation is very simple, fast, and getter-free, and is as effective as glass encapsulation. Use

Articles

Fuel cells have the potential to reduce the nation’s energy use through increased energy conversion efficiency and dependence on imported petroleum by the use of hydrogen from renewable resources.

Proton exchange membrane (PEM) fuel cells operate at relatively low temperatures and are composed of two electrodes and a conductive elecrolyte.

Advances in the electrochemical conversion of water to and from hydrogen and oxygen have principally been achieved through the development of new materials and by understanding the mechanisms of the degradation of proton exchange membrane fuel cells (PEMFC) during operation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service