Skip to Content
Merck
All Photos(1)

Key Documents

479780

Sigma-Aldrich

Diethylenetriamine, polymer-bound

200-400 mesh, extent of labeling: 2.5-3.0 mmol/g loading, 2 % cross-linked

Synonym(s):

PS-DETA

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

crosslinking

2 % cross-linked

reaction suitability

reaction type: solution phase peptide synthesis

extent of labeling

2.5-3.0 mmol/g loading

particle size

200-400 mesh

functional group

amine

SMILES string

NCCNCCN.C=Cc1ccccc1.C=Cc2ccc(C=C)cc2

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service