Skip to Content
Merck
All Photos(1)

Key Documents

409774

Sigma-Aldrich

Trimethylolpropane ethoxylate

average Mn ~450

Synonym(s):

3 arm PEG, 3 arm PEO, Poly(ethylene oxide), 3-arm, hydroxy terminated, Polyethylene glycol trimethylolpropane ether, Polyethylene glycol trimethylolpropane triether, Trimethylolpropane ethoxylate (20/3 EO/OH), Trimethylolpropane ethoxylate (4.6 EO/OH), Trimethylolpropane ethoxylate (4/15 EO/OH)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H4O)n(C2H4O)n(C2H4O)nC6H14O
CAS Number:
MDL number:
UNSPSC Code:
12162002

mol wt

average Mn ~450

refractive index

n20/D 1.472

density

1.114 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

Application

Trimethylolpropane ethoxylate (TMPE) can be dissolved with lithium trifluoromethanesulfonate (LiCF3SO3) to form an electrolyte. These electrolytes can act as charge transferring mediums for light emitting electrochemical cells and devices.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

444.2 °F - closed cup

Flash Point(C)

229 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Small-molecule light-emitting electrochemical cells: evidence for in situ electrochemical doping and functional operation
Tang S, et al.
Chemical Communications (Cambridge, England), 49(43), 4926-4928 (2013)
Identifying key properties of electrolytes for light-emitting electrochemical cells
Tang S, et al.
Chemistry of Materials, 26(17), 5083-5088 (2014)
Quest for an appropriate electrolyte for high-performance light-emitting electrochemical cells
Tang S and Edman L
The Journal of Physical Chemistry Letters, 1(18), 2727-2732 (2010)
White light from a single-emitter light-emitting electrochemical cell
Tang S, et al.
Journal of the American Chemical Society, 135(9), 3647-3652 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service