323616
3′-Deoxythymidine
98%
Synonym(s):
2′,3′-Dideoxythymidine, DDT
About This Item
Recommended Products
Assay
98%
optical activity
[α]19/D +20.0°, c = 0.6 in H2O
mp
155-156 °C (lit.)
functional group
ether
hydroxyl
SMILES string
CC1=CN([C@H]2CC[C@@H](CO)O2)C(=O)NC1=O
InChI
1S/C10H14N2O4/c1-6-4-12(10(15)11-9(6)14)8-3-2-7(5-13)16-8/h4,7-8,13H,2-3,5H2,1H3,(H,11,14,15)/t7-,8+/m0/s1
InChI key
XKKCQTLDIPIRQD-JGVFFNPUSA-N
Gene Information
mouse ... Esr1(13982)
Looking for similar products? Visit Product Comparison Guide
Application
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Global Trade Item Number
SKU | GTIN |
---|---|
323616-25MG | |
323616-100MG |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service