Skip to Content
Merck
All Photos(2)

Key Documents

283266

Sigma-Aldrich

4-Sulfophthalic acid solution

50 wt. % in H2O, technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO3SC6H3-1,2-(CO2H)2
CAS Number:
Molecular Weight:
246.19
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

concentration

50 wt. % in H2O

refractive index

n20/D 1.447

density

1.292 g/mL at 25 °C

SMILES string

OC(=O)c1ccc(cc1C(O)=O)S(O)(=O)=O

InChI

1S/C8H6O7S/c9-7(10)5-2-1-4(16(13,14)15)3-6(5)8(11)12/h1-3H,(H,9,10)(H,11,12)(H,13,14,15)

InChI key

WNKQDGLSQUASME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

major contaminant is 3-sulfophthalic acid

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junquan Meng et al.
ACS applied materials & interfaces, 11(31), 28461-28468 (2019-07-12)
To solve the pore-wetting problems of membrane distillation, we developed a series of three-layer composite pervaporation (PV) membranes that could be applied in direct contact mode. Specifically, a porous poly(vinylidene fluoride) (PVDF) layer was placed on top of a polytetrafluoroethylene
En-Cui Yang et al.
Dalton transactions (Cambridge, England : 2003), 44(7), 3190-3199 (2015-01-13)
Four new 4-amino-1,2,4-triazole (atr)-based coordination polymers, {[Cu2(atr)(H2O)(μ-OH)2(pa)]·H2O}n (), {[Cu3(atr)2(H2O)2(μ-OH)2(npa)2]·2H2O}n (), {[Cu3(atr)5(dca)(μ-OH)(ClO4)2](ClO4)2}n () and {[Cu3(atr)2(H2O)(μ3-OH)(μ-OH)2(spa)]·1.5H2O}n () (pa(2-) = phthalate, npa(2-) = 3-nitrophthalate, dca(-) = dicyanamide and spa(3-) = 4-sulfophthalate), were successfully obtained by varying the carboxylate- and cyanide-modified magnetic bridges. Structural

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service