Skip to Content
Merck
All Photos(1)

Key Documents

261084

Sigma-Aldrich

Erbium

ingot, 99.9% trace rare earth metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Er
CAS Number:
Molecular Weight:
167.26
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace rare earth metals basis

form

ingot

reaction suitability

reagent type: catalyst
core: erbium

resistivity

86 μΩ-cm, 20°C

bp

2868 °C (lit.)

mp

1529 °C (lit.)

density

9.062 g/mL at 25 °C (lit.)

SMILES string

[Er]

InChI

1S/Er

InChI key

UYAHIZSMUZPPFV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Etienne Pelletier et al.
Optics express, 20(25), 27456-27464 (2012-12-25)
We demonstrate the generation of 22.6 μJ of combined energy at 3 μm for sub-300fs pulses at a repetition rate of 1 kHz using a LGSe optical parametric amplifier (OPA). The LGSe OPA is pumped by the 140-fs 1.6 μm
Guillaume Le Cocq et al.
Optics express, 20(24), 27051-27061 (2012-11-29)
Numerical and experimental study of a Few-Mode (FM) Erbium Doped Fiber Amplifier (EDFA) suitable for mode division multiplexing (MDM) is reported. Based on numerical simulations, a Few-Mode Erbium Doped Fiber (FM-EDF) has been designed to amplify four mode groups and
Hans-Martin Eiter et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(1), 64-69 (2012-12-19)
Charge and spin density waves, periodic modulations of the electron, and magnetization densities, respectively, are among the most abundant and nontrivial low-temperature ordered phases in condensed matter. The ordering direction is widely believed to result from the Fermi surface topology.
Chujun Zhao et al.
Optics express, 20(25), 27888-27895 (2012-12-25)
Based on the open-aperture Z-scan measurement, we firstly uncovered the saturable absorption property of the topological insulator (TI): Bi2Se3. A high absolute modulation depth up to 98% and a saturation intensity of 0.49 GWcm(-2) were identified. By incorporating this novel
Junghan Lee et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(1), 96-103 (2012-12-13)
Multimodal nanoparticles have been extensively studied for target-specific imaging and therapy of various diseases, including cancer. In this study, radiolabeled arginine-glycine-aspartic acid (RGD)-functionalized Er(3+)/Yb(3+) co-doped NaGdF(4) upconversion nanophosphors (UCNPs) were synthesized and evaluated as a multimodal PET/MR/optical probe with tumor

Articles

A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.

The application of magnetism and magnetic materials pervades our modern civilization in the form of electrical power, communications and information storage.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

The unique properties of the rare-earth elements and their alloys have brought them from relative obscurity to high profile use in common hightech applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service