246018
2,3,5-Triiodobenzoic acid
technical grade
Synonym(s):
TIBA
About This Item
Recommended Products
grade
technical grade
mp
220-222 °C (lit.)
solubility
methanol: 5%, hazy, orange to very deep orange (light)
SMILES string
OC(=O)c1cc(I)cc(I)c1I
InChI
1S/C7H3I3O2/c8-3-1-4(7(11)12)6(10)5(9)2-3/h1-2H,(H,11,12)
InChI key
ZMZGFLUUZLELNE-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service