Skip to Content
Merck
All Photos(1)

Key Documents

MABF320

Sigma-Aldrich

Anti-CD45 (mouse) Antibody, FITC, clone 30-F11

clone 30-F11, from rat, FITC conjugate

Synonym(s):

Receptor-type tyrosine-protein phosphatase C, Leukocyte common antigen, L-CA, T200, CD45

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

biological source

rat

conjugate

FITC conjugate

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

30-F11, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PTPRC(5788)

General description

CD45, also known as PTPRC, is a receptor-tyrosine phosphatase required for antigen-induced signaling in T-cells and B-cells. CD 45 belongs to the protein-tyrosine phosphatase family, receptor class 1/6 subfamily.It also contains 2 fibronectin type-III domains and 2 tyrosine-protein phosphatase domains. CD45 has been shown to play an important role in T-cell activation through the antigen receptor. CD45 also is suggested to act as a positive regulator of T-cell coactivation upon binding to DPP4. The first PTPase domain has enzymatic activity, while the second one is suggested to affect the substrate specificity of the first domain. Upon T-cell activation, CD45 recruits and dephosphorylates SKAP1 and FYN. Furthermore, it has been shown to dephosphorylate LYN, and modulate LYN activity. Interestingly, CD45 has been associated with several Src-family tyrosine kinases, which are inhibited by tyrosine phosphorylation in their c-termini, suggesting a mechanism for signaling by CD45. CD45 has been shown to bind with GANAB and PRKCSH. Additionally, it interacts with SKAP1 and DPP4, whereas the interaction is enhanced in a interleukin-12-dependent manner in activated lymphocytes. Mutations in CD45 have been associated with Multiple Sclerosis (MS) and severe combined immunodeficiency (SCID), a genetically and clinically heterogeneous group of rare congenital disorders characterized by impairment of both humoral and cell-mediated immunity, leukopenia, and low or absent antibody levels.

Immunogen

Mouse thymus or spleen

Application

Anti-CD45 (mouse) Antibody, FITC, clone 30-F11 is a highly specific rat monoclonal antibody, that targets CD45 & has been tested in Flow Cytometry, IHC, western blotting & IP.
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors
Western Blotting Analysis: A representative lot detected human CD45 in Western Blotting. (Tsuboi S and Fukuda M. 1998)

Immunohistochemistry Analysis: A representative lot detected human CD45 in Immunohistochemistry. (Simon DI, et al. 2000; Cornet A, et al. 2001)

Immunoprecipitation Analysis: A representative lot detected human CD45 in Immunoprecipitation. (Ledbetter JA, et al. 1979)

Quality

The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC and unconjugated antibody.

Flow Cytometry Analysis: 0.25 μg of this antibody detected CD45 in one million C57BL/6 mouse splenocytes.

Target description

145 kDa calculated

Physical form

Affinity
Purified rat conjugate antibody in buffer containing PBS (pH 7.2) with up to 0.1% sodium azide.

Storage and Stability

The antibody solution should be stored undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Binhui Zhou et al.
Frontiers in immunology, 11, 607442-607442 (2021-01-26)
Zdhhc family genes are composed of 24 members that regulate palmitoylation, a post-translational modification process for proteins. Mutations in genes that alter palmitoylation or de-palmitoylation could result in neurodegenerative diseases and inflammatory disorders. In this study, we found that Zdhhc2
Overexpression of branched O-linked oligosaccharides on T cell surface glycoproteins impairs hu?moral immune responses in transgenic mice.
Tsuboi, S and Fukuda, M
The Journal of Biological Chemistry, 273, 30680-30687 (1998)
D I Simon et al.
The Journal of clinical investigation, 105(3), 293-300 (2000-02-17)
Inflammation plays an essential role in the initiation and progression of atherosclerosis, but its role in vascular repair after mechanical arterial injury (i.e., percutaneous transluminal coronary angioplasty, PTCA) is unknown. In animal models of vascular injury, leukocytes are recruited as
A Cornet et al.
Proceedings of the National Academy of Sciences of the United States of America, 98(23), 13306-13311 (2001-11-01)
Early pathological manifestations of Crohn's disease (CD) include vascular disruption, T cell infiltration of nerve plexi, neuronal degeneration, and induction of T helper 1 cytokine responses. This study demonstrates that disruption of the enteric glial cell network in CD patients
Xenogeneic monoclonal antibodies to mouse lymphoid differentiation antigens.
J A Ledbetter et al.
Immunological reviews, 47, 63-90 (1979-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service